Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127566.1 | NP_001121038.1 | 5 - 6 | 700 | 77 | |
GenBank mRNA | BC161896.1 | - | 5 - 6 | 700 | 7319876 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |