Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009663.1 | NP_001009663.1 | 1 - 2 | 49 | 54 | |
GenBank mRNA | BC088300.1 | - | 1 - 2 | 49 | 7323342 | 54 |
FQ209415.1 | - | 1 - 2 | 94 | 54 | ||
FQ211289.1 | - | 1 - 2 | 64 | 54 | ||
FQ218294.1 | - | 1 - 2 | 69 | 54 | ||
FQ218398.1 | - | 1 - 2 | 58 | 54 | ||
FQ219289.1 | - | 1 - 2 | 123 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |