Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106743.1 | NP_001100213.1 | 9 - 10 | 1988 | 74 | |
XM_006240323.2 | XP_006240385.1 | 9 - 10 | 2462 | 74 | ||
XM_006240324.2 | XP_006240386.1 | 9 - 10 | 2458 | 74 | ||
XM_006240325.2 | XP_006240387.1 | 9 - 10 | 2403 | 74 | ||
XM_008764752.1 | XP_008762974.1 | 6 - 7 | 1375 | 74 | ||
XM_008764753.1 | XP_008762975.1 | 6 - 7 | 1551 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |