Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011983.1 | NP_001011983.1 | 8 - 9 | 940 | 63 | |
GenBank mRNA | BC079342.1 | - | 8 - 9 | 940 | 7106851 | 63 |
FQ228932.1 | - | 7 - 8 | 1297 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |