Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034929.1 | NP_001030101.1 | 3 - 4 | 686 | 87 | |
XM_006240283.2 | XP_006240345.1 | 3 - 4 | 990 | 87 | ||
XM_008764784.1 | XP_008763006.1 | 2 - 3 | 1119 | 87 | ||
GenBank mRNA | BC105849.1 | - | 3 - 4 | 686 | 7457541 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |