Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044235.2 | NP_001037700.1 | 5 - 6 | 393 | 95 | |
GenBank mRNA | AB256045.1 | - | 5 - 6 | 396 | 95 | |
AM258960.1 | - | 5 - 6 | 396 | 95 | ||
FQ222819.1 | - | 5 - 6 | 394 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |