Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108050.2 | NP_001101520.2 | 2 - 3 | 664 | 108 | |
NM_001271174.1 | NP_001258103.1 | 2 - 3 | 922 | 108 | ||
GenBank mRNA | CA513249.1 | - | 2 - 3 | 546 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |