Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005888.1 | NP_001005888.1 | 15 - 16 | 1826 | 136 | |
XM_008764799.1 | XP_008763021.1 | 15 - 16 | 2262 | 136 | ||
GenBank mRNA | AY386370.1 | - | 15 - 16 | 1826 | 136 | |
FQ223346.1 | - | 15 - 16 | 1845 | 136 | ||
FQ228993.1 | - | 15 - 16 | 1845 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |