Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005888.1 | NP_001005888.1 | 4 - 5 | 438 | 68 | |
XM_008764799.1 | XP_008763021.1 | 4 - 5 | 874 | 68 | ||
GenBank mRNA | AY386370.1 | - | 4 - 5 | 438 | 68 | |
FQ223346.1 | - | 4 - 5 | 457 | 68 | ||
FQ228993.1 | - | 4 - 5 | 457 | 68 |
Gene Symbol: LOC100910182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764910.1 | XP_008763132.1 | 3 - 4 | 308 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |