Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078620.1 | NP_511175.1 | 2 - 3 | 2618 | 105 | |
GenBank mRNA | U53420.1 | - | 2 - 3 | 2618 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |