Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc8a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078620.1 | NP_511175.1 | 3 - 4 | 2721 | 64 | |
GenBank mRNA | U53420.1 | - | 3 - 4 | 2721 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Rattus norvegicus Probe spans exons |