Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163153.1 | NP_001156625.1 | 3 - 4 | 352 | 214 | |
GenBank mRNA | FQ210472.1 | - | 3 - 4 | 345 | 214 | |
FQ211105.1 | - | 3 - 4 | 349 | 214 | ||
FQ224072.1 | - | 3 - 4 | 340 | 214 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |