Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109442.1 | NP_001102912.1 | 2 - 3 | 405 | 100 | |
XM_008764848.1 | XP_008763070.1 | 1 - 2 | 89 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |