Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109442.1 | NP_001102912.1 | 3 - 4 | 484 | 96 | |
XM_006240290.2 | XP_006240352.1 | 2 - 3 | 186 | 96 | ||
XM_008764848.1 | XP_008763070.1 | 2 - 3 | 168 | 96 | ||
GenBank mRNA | BC166864.1 | - | 2 - 3 | 101 | 8367571 | 96 |
FQ224144.1 | - | 2 - 3 | 191 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |