Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053467.1 | NP_445919.1 | 3 - 4 | 416 | 70 | |
GenBank mRNA | AJ004912.1 | - | 3 - 4 | 416 | 70 | |
FQ212056.1 | - | 3 - 4 | 450 | 70 | ||
X97443.1 | - | 3 - 4 | 368 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |