Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108053.1 | NP_001101523.1 | 1 - 2 | 195 | 142 | |
GenBank mRNA | BC168869.1 | - | 1 - 2 | 112 | 7104604 | 142 |
FQ211938.1 | - | 1 - 2 | 181 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |