Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021655.2 | NP_067687.2 | 1 - 2 | 194 | 64 | |
GenBank mRNA | AF145445.1 | - | 1 - 2 | 56 | 64 | |
BC087703.1 | - | 1 - 2 | 174 | 7313842 | 64 | |
CB743841.1 | - | 1 - 2 | 250 | 64 | ||
X06832.1 | - | 1 - 2 | 179 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |