Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021655.2 | NP_067687.2 | 6 - 7 | 1001 | 90 | |
GenBank mRNA | AF145445.1 | - | 6 - 7 | 854 | 90 | |
BC087703.1 | - | 6 - 7 | 981 | 7313842 | 90 | |
X06832.1 | - | 6 - 7 | 989 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |