Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014043.1 | NP_001014065.1 | 1 - 2 | 214 | 67 | |
GenBank mRNA | BC085803.1 | - | 1 - 2 | 214 | 7111151 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |