Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025271.1 | NP_001020442.1 | 6 - 7 | 1748 | 87 | |
GenBank mRNA | AF036335.1 | - | 2 - 3 | 277 | 87 | |
BC098731.1 | - | 6 - 7 | 1748 | 7384804 | 87 | |
FQ227268.1 | - | 2 - 3 | 213 | 87 | ||
FQ229853.1 | - | 2 - 3 | 290 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |