Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025271.1 | NP_001020442.1 | 8 - 9 | 1919 | 81 | |
GenBank mRNA | AF036335.1 | - | 4 - 5 | 449 | 81 | |
BC098731.1 | - | 8 - 9 | 1919 | 7384804 | 81 | |
FQ227268.1 | - | 4 - 5 | 384 | 81 | ||
FQ229853.1 | - | 4 - 5 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |