Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017230.2 | NP_058926.1 | 3 - 4 | 389 | 67 | |
GenBank mRNA | D88034.1 | - | 3 - 4 | 390 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |