Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107914.1 | NP_001101384.1 | 11 - 12 | 2156 | 79 | |
XM_006238934.1 | XP_006238996.1 | 12 - 13 | 2264 | 79 | ||
XM_006238935.2 | XP_006238997.1 | 12 - 13 | 2627 | 79 | ||
XM_006238936.1 | XP_006238998.1 | 12 - 13 | 2264 | 79 | ||
XM_006238937.1 | XP_006238999.1 | 12 - 13 | 2264 | 79 | ||
XM_006238938.1 | XP_006239000.1 | 11 - 12 | 2099 | 79 | ||
XM_006238939.1 | XP_006239001.1 | 11 - 12 | 2099 | 79 | ||
XM_008764141.1 | XP_008762363.1 | 12 - 13 | 2264 | 79 | ||
XM_008764142.1 | XP_008762364.1 | 11 - 12 | 2099 | 79 | ||
XM_008764143.1 | XP_008762365.1 | 11 - 12 | 2099 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |