Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011977.1 | NP_001011977.1 | 3 - 4 | 308 | 65 | |
XM_006238860.1 | XP_006238922.1 | 3 - 4 | 480 | 65 | ||
XM_006238861.2 | XP_006238923.1 | 2 - 3 | 241 | 65 | ||
GenBank mRNA | BC078990.1 | - | 3 - 4 | 308 | 7112575 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |