Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031607.1 | NP_113795.1 | 3 | 66 | ||
GenBank mRNA | BC089930.1 | - | 1 | 66 | ||
BC169050.1 | - | 3 | 7103781 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |