Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053543.2 | NP_445995.2 | 5 - 6 | 1970 | 89 | |
XM_006238908.1 | XP_006238970.1 | 5 - 6 | 1679 | 89 | ||
XM_006238909.1 | XP_006238971.1 | 5 - 6 | 1740 | 89 | ||
GenBank mRNA | BC087000.1 | - | 5 - 6 | 1970 | 7190905 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |