Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013216.2 | NP_001013234.2 | 6 - 7 | 1315 | 56 | |
XM_006239469.2 | XP_006239531.1 | 6 - 7 | 1324 | 56 | ||
XM_008764367.1 | XP_008762589.1 | 6 - 7 | 1353 | 56 | ||
GenBank mRNA | BC091214.1 | - | 5 - 6 | 1914 | 7383542 | 56 |
FQ211844.1 | - | 6 - 7 | 1342 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |