Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Matn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006979.1 | NP_001006980.1 | 2 - 3 | 468 | 94 | |
GenBank mRNA | BC083869.1 | - | 2 - 3 | 468 | 7128388 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |