Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083122.1 | NP_001076591.1 | 5 - 6 | 804 | 78 | |
NM_012890.2 | NP_037022.1 | 4 - 5 | 743 | 78 | ||
GenBank mRNA | BC061997.1 | - | 4 - 5 | 746 | 78 | |
BC168237.1 | - | 5 - 6 | 797 | 7938890 | 78 | |
U50927.1 | - | 4 - 5 | 743 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons |