Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031648.1 | NP_001026818.1 | 1 - 2 | 332 | 84 | |
GenBank mRNA | BC098936.1 | - | 1 - 2 | 332 | 7446346 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |