Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750087.3 | XP_003750135.1 | 3 - 4 | 1587 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |