Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764190.1 | XP_008762412.1 | 5 - 6 | 830 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |