Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764190.1 | XP_008762412.1 | 6 - 7 | 964 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |