Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134689.1 | NP_001128161.1 | 1 - 2 | 53 | 107 | |
GenBank mRNA | CB711403.1 | - | 1 - 2 | 63 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |