Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134689.1 | NP_001128161.1 | 2 - 3 | 239 | 110 | |
XM_008764171.1 | XP_008762393.1 | 2 - 3 | 3034 | 110 | ||
GenBank mRNA | BC168155.1 | - | 1 - 2 | 616 | 8362701 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |