Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108687.1 | NP_001102157.1 | 2 - 3 | 623 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |