Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tp73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108696.1 | NP_001102166.1 | 6 - 7 | 874 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Rattus norvegicus Probe spans exons |