Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239327.2 | XP_006239389.1 | 1 - 2 | 721 | 94 | |
XM_008764305.1 | XP_008762527.1 | 1 - 2 | 721 | 94 | ||
XM_008764306.1 | XP_008762528.1 | 1 - 2 | 721 | 94 | ||
XM_238429.8 | XP_238429.5 | 1 - 2 | 721 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |