Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764306.1 | XP_008762528.1 | 5 - 6 | 1722 | 89 | |
XM_238429.8 | XP_238429.5 | 5 - 6 | 1581 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |