Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239282.2 | XP_006239344.1 | 2 - 3 | 534 | 62 | |
XM_006239283.2 | XP_006239345.1 | 2 - 3 | 192 | 62 | ||
XM_008764300.1 | XP_008762522.1 | 3 - 4 | 522 | 62 | ||
XM_008764301.1 | XP_008762523.1 | 3 - 4 | 525 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |