Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100538.1 | NP_001094008.1 | 14 - 15 | 2218 | 99 | |
XM_006238798.1 | XP_006238860.1 | 14 - 15 | 2806 | 99 | ||
XM_006238799.1 | XP_006238861.1 | 14 - 15 | 2806 | 99 | ||
XM_006238800.1 | XP_006238862.1 | 13 - 14 | 2719 | 99 | ||
XM_006238801.1 | XP_006238863.1 | 13 - 14 | 2671 | 99 | ||
GenBank mRNA | BC088337.1 | - | 9 - 10 | 1165 | 99 | |
BC166452.1 | - | 14 - 15 | 2223 | 8372968 | 99 |
Gene Symbol: RGD1562451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 2240 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |