Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100538.1 | NP_001094008.1 | 8 - 9 | 1610 | 88 | |
XM_006238798.1 | XP_006238860.1 | 8 - 9 | 2150 | 88 | ||
XM_006238799.1 | XP_006238861.1 | 8 - 9 | 2150 | 88 | ||
XM_006238800.1 | XP_006238862.1 | 8 - 9 | 2150 | 88 | ||
XM_006238801.1 | XP_006238863.1 | 8 - 9 | 2150 | 88 | ||
GenBank mRNA | BC088337.1 | - | 3 - 4 | 509 | 88 | |
BC166452.1 | - | 8 - 9 | 1615 | 8372968 | 88 |
Gene Symbol: RGD1562451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 1584 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |