Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764322.1 | XP_008762544.1 | 8 - 9 | 1653 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |