Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127591.1 | NP_001121063.1 | 9 - 10 | 1459 | 54 | |
XM_008764385.1 | XP_008762607.1 | 9 - 10 | 2078 | 54 | ||
XM_008764386.1 | XP_008762608.1 | 9 - 10 | 2245 | 54 | ||
XM_008764387.1 | XP_008762609.1 | 9 - 10 | 1632 | 54 | ||
GenBank mRNA | BC166429.1 | - | 9 - 10 | 1459 | 7128351 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |