Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tardbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011979.2 | NP_001011979.1 | 1 - 2 | 91 | 96 | |
XM_006239382.2 | XP_006239444.1 | 1 - 2 | 144 | 96 | ||
XM_006239384.2 | XP_006239446.1 | 1 - 2 | 121 | 96 | ||
XM_006239385.2 | XP_006239447.1 | 1 - 2 | 122 | 96 | ||
XM_006239386.2 | XP_006239448.1 | 1 - 2 | 126 | 96 | ||
XM_008764234.1 | XP_008762456.1 | 1 - 2 | 123 | 96 | ||
XM_008764235.1 | XP_008762457.1 | 1 - 2 | 121 | 96 | ||
XM_008764236.1 | XP_008762458.1 | 1 - 2 | 121 | 96 | ||
GenBank mRNA | AI502405.1 | - | 1 - 2 | 102 | 96 | |
BC083752.1 | - | 1 - 2 | 80 | 7192591 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |