Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_213954.8 | XP_213954.4 | 19 - 20 | 3739 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |