Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsdhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009399.1 | NP_001009399.1 | 4 - 5 | 492 | 69 | |
XM_006229515.2 | XP_006229577.1 | 4 - 5 | 507 | 69 | ||
GenBank mRNA | BC087626.1 | - | 4 - 5 | 492 | 7317527 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |