Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhatl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106868.1 | NP_001100338.1 | 2 - 3 | 273 | 111 | |
XM_006244090.2 | XP_006244152.1 | 2 - 3 | 1175 | 111 | ||
XM_006244091.2 | XP_006244153.1 | 2 - 3 | 354 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |