Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233537.2 | XP_006233599.1 | 4 - 5 | 705 | 82 | |
XM_227798.8 | XP_227798.5 | 4 - 5 | 707 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |