Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017449.1 | NP_001017449.1 | 3 - 4 | 577 | 91 | |
XM_006233214.1 | XP_006233276.1 | 3 - 4 | 390 | 91 | ||
XM_008761454.1 | XP_008759676.1 | 2 - 3 | 438 | 91 | ||
XM_008761455.1 | XP_008759677.1 | 2 - 3 | 439 | 91 | ||
GenBank mRNA | BC092598.1 | - | 3 - 4 | 577 | 7321341 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |